GC Analysis of 1,2-Alkyldiol Enantiomers (O-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-TA
Materials
analytical column
standard
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
80 °C, 5 °C/min to 130 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample
various alkyldiol enantiomers (O-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC