Merck
GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX B-DA
GC Analysis of 1,2,3,4-Tetrahydro-1-Naphthylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX™ B-DA suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-DA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
1,2,3,4-Tetrahydronaphthalene
analytical standard
1,2,3,4-Tetrahydronaphthalene
anhydrous, 99%
CONDITIONS
column
Astec CHIRALDEX B-DA, 30 m x 0.25 mm I.D., 0.12 μm (72023AST)
oven
180 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: 1,2,3,4-tetrahydro-1-naphthylamine enantiomers (N-acetyl derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC