Merck
US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
CONDITIONS
column
SLB-5ms, 20 m × 0.18 mm I.D., 0.18 μm (28564-U)
oven
45 °C, 86 °C/min to 130 °C (1.73 min), 20 °C/min to 180 °C, 12 °C/min to 240 °C, 20 °C/min to 330 °C (2 min)
inj. temp.
275 °C
MSD interface
330 °C
scan range
m/z = 40-450
carrier gas
helium, 1.4 mL/min, constant
injection
1 μL splitless (0.75 min)
liner
2 mm I.D. fast FocusLiner liner with taper
sample
109 component semivolatile standard (each analyte at 2 ppm), plus 3 surrogate and 3 internal standard compounds (each at 5 ppm) in ethyl acetate
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany