- Prevalence, mechanisms and genetic relatedness of the human pathogenic fungus Aspergillus fumigatus exhibiting resistance to medical azoles in the environment of Taiwan.
Prevalence, mechanisms and genetic relatedness of the human pathogenic fungus Aspergillus fumigatus exhibiting resistance to medical azoles in the environment of Taiwan.
Emerging azole resistance in Aspergillus fumigatus poses a serious threat to human health. This nationwide surveillance study investigated the prevalence and molecular characteristics of azole-resistant A. fumigatus environmental isolates in Taiwan, an island country with increasing use of azole fungicides. Of the 2760 air and soil samples screened from 2014 to 2016, 451 A. fumigatus isolates were recovered from 266 samples and 34 isolates from 29 samples displayed resistance to medical azoles (itraconazole, voriconazole or posaconazole). The resistance prevalence was 10.9% and 7.5% in A. fumigatus-positive samples and isolates respectively. Most (29, 85.3%) azole-resistant isolates harboured TR