Merck
  • Simple mixed Fe-Zn catalysts for the Suzuki couplings of tetraarylborates with benzyl halides and 2-halopyridines.
Simple mixed Fe-Zn catalysts for the Suzuki couplings of tetraarylborates with benzyl halides and 2-halopyridines.
Chemical communications (Cambridge, England) (2009-10-21)
Robin B Bedford, Mark A Hall, George R Hodges, Michael Huwe, Mark C Wilkinson
ABSTRACT
Employing co-catalytic zinc reagents facilitates the iron-catalysed Suzuki cross-coupling of tetraarylborates with both benzyl and 2-heteroaryl halides.
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer