Merck
Gold-Doping of Double-Crown Pd Nanoclusters.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-10-17)
Jishi Chen, Liren Liu, Xu Liu, Lingwen Liao, Shengli Zhuang, Shiming Zhou, Jinlong Yang, Zhikun Wu
ABSTRACT
Double-crown Ni, Pd, or Pt nanoclusters have attracted extensive interests due to their aesthetic structure and intriguing properties. However, their doping by other metals remains unknown until now. Herein, Pd
Sigma-Aldrich
Phenylethyl mercaptan, ≥99%, FG