Merck
Effect of convection and B
Journal of magnetic resonance (San Diego, Calif. : 1997) (2017-09-20)
B Kharkov, X Duan, J W Canary, A Jerschow
ABSTRACT
Nuclear spin singlet lifetimes can often exceed the T
Sigma-Aldrich
Dimethyl maleate, 96%
Pricing and availability is not currently available.