Merck
  • Safflor yellow B reduces hypoxia-mediated vasoconstriction by regulating endothelial micro ribonucleic acid/nitric oxide synthase signaling.
Safflor yellow B reduces hypoxia-mediated vasoconstriction by regulating endothelial micro ribonucleic acid/nitric oxide synthase signaling.
Oncotarget (2017-12-08)
Chaoyun Wang, Ying Yang, Miao Li, Xin Liu, Qiaoyun Wang, Wenyu Xin, Hongliu Sun, Qingyin Zheng
ABSTRACT
Hypoxia-induced generation of vasoconstrictors reduces cerebral blood flow (CBF) while nitric oxide (NO) synthase (NOS) and microRNAs (miRNA) in endothelial cells (ECs) suppress vasoconstriction. Safflor yellow B (SYB), a natural plant compound, previously attenuated angiotensin II-mediated injury of ECs and maintained endothelial function. This study investigated the putative involvement of NOS and miRNAs in SYB-mediated resistance to hypoxia-induced vasoconstriction. In vivo, chronic hypoxia was induced in rats, and SYB was administered intravenously. In vitro, rat primary aortic ECs were cultured under oxygen and glucose deprivation. After treatment with anti-microR-199a, as well as the NOS inhibitor, N(G)-nitro-L-arginine methyl ester, SYB, or both, cell viability, NO and peroxynitrite (ONOO-) levels, NOS expression, and miRNA levels were evaluated. SYB significantly alleviated hypoxia-mediated vasoconstriction and increased CBF endothelium-dependently. SYB upregulated miR-199a, increased EC viability, decreased endothelin-1 (ET-1) levels, inhibited protein kinase C (PKC) activity, and suppressed hypoxia inducible factor-1α (HIF-1α) expression. Furthermore, the SYB-mediated reduction of inducible NOS reduced ONOO- levels. In addition, SYB downregulated miR-138 and, thereby, enhanced S100A1 and endothelial NOS activity. Hypoxia-mediated regulation of miR-138 and miR-199a inhibited endothelial NOS expression and activation, which triggered ET-1 release and vasoconstriction. Therefore, SYB treatment reduced hypoxia-induced vasoconstriction through miR-199a/endothelial NOS signaling.
Sigma-Aldrich
Anti-BAX antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-S100 A1, N-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-BCL-2 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-INOS antibody produced in rabbit, affinity isolated antibody