Merck
  • Single-provenance mature conifers show higher non-structural carbohydrate storage and reduced growth in a drier location.
Single-provenance mature conifers show higher non-structural carbohydrate storage and reduced growth in a drier location.
Tree physiology (2017-05-27)
Frida I Piper, Alex Fajardo, Günter Hoch
ABSTRACT
Since growth is more sensitive to drought than photosynthesis, trees inhabiting dry regions are expected to exhibit higher carbohydrate storage and less growth than their conspecifics from more humid regions. However, the same pattern can be the result of different genotypes inhabiting contrasting humidity conditions. To test if reduced growth and high carbohydrate storage are environmentally driven by drought, we examined the growth and non-structural carbohydrate (NSC) concentrations in single-provenance stands of mature trees of Pinus contorta Douglas and Pinus ponderosa Douglas ex C. Lawson planted at contrasting humidity conditions (900 versus 300 mm of annual precipitation) in Patagonia, Chile. Individual tree growth was measured for each species and at each location as mean basal area increment of the last 10 years (BAI10), annual shoot elongation for the period 2011-14, and needle length for 2013 and 2014 cohorts. Additionally, needle, branch, stem sapwood and roots were collected from each sampled tree to determine soluble sugars, starch and total NSC concentrations. The two species showed lower mean BAI10 and 2013 needle length in the dry site; P. ponderosa also had lower annual shoot extension for 2011 and 2014, and lower 2014 needle length, in the dry than in the mesic site. By contrast, NSC concentrations of all woody tissues for both species were either similar or higher in the dry site when compared with the mesic site. Patterns of starch and sugars were substantially different: starch concentrations were similar between sites except for roots of P. ponderosa, which were higher in the dry site, while sugar concentrations of all woody tissues in both species were higher in the dry site. Overall, our study provides evidence that reduced growth along with carbon (C) accumulation is an environmentally driven response to drought. Furthermore, the significant accumulation of low-molecular weight sugars in the dry site is compatible with a prioritized C allocation for osmoregulation. However, since this accumulation did not come at the expense of reduced starch, it is unlikely that growth was limited by C supply in the dry site.
Sigma-Aldrich
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)
Pricing and availability is not currently available.
Supelco
Glucose Assay Reagent, for use with enzymatic assay kits GAHK20, SCA20, FA20, SA20
Pricing and availability is not currently available.
Sigma-Aldrich
Invertase from baker′s yeast (S. cerevisiae), Grade VII, ≥300 units/mg solid
Pricing and availability is not currently available.