Merck
  • Control of Cell Shape, Neurite Outgrowth, and Migration by a Nogo-A/HSPG Interaction.
Control of Cell Shape, Neurite Outgrowth, and Migration by a Nogo-A/HSPG Interaction.
Developmental cell (2017-09-26)
Anissa Kempf, Enrica Boda, Jessica C F Kwok, Rafael Fritz, Valentina Grande, Andrea M Kaelin, Zorica Ristic, Andre Schmandke, Antonio Schmandke, Bjoern Tews, James W Fawcett, Olivier Pertz, Annalisa Buffo, Martin E Schwab
ABSTRACT
Heparan sulfate proteoglycans (HSPGs) critically modulate adhesion-, growth-, and migration-related processes. Here, we show that the transmembrane protein, Nogo-A, inhibits neurite outgrowth and cell spreading in neurons and Nogo-A-responsive cell lines via HSPGs. The extracellular, active 180 amino acid Nogo-A region, named Nogo-A-Δ20, binds to heparin and brain-derived heparan sulfate glycosaminoglycans (GAGs) but not to the closely related chondroitin sulfate GAGs. HSPGs are required for Nogo-A-Δ20-induced inhibition of adhesion, cell spreading, and neurite outgrowth, as well as for RhoA activation. Surprisingly, we show that Nogo-A-Δ20 can act via HSPGs independently of its receptor, Sphingosine-1-Phosphate receptor 2 (S1PR2). We thereby identify the HSPG family members syndecan-3 and syndecan-4 as functional receptors for Nogo-A-Δ20. Finally, we show in explant cultures ex vivo that Nogo-A-Δ20 promotes the migration of neuroblasts via HSPGs but not S1PR2.
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
Heparan sulfate sodium salt from bovine kidney
Sigma-Aldrich
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human SDC3