Merck
  • Formation and rheological behavior of wormlike micelles in a catanionic system of fluoroacetic acid and tetradecyldimethylaminoxide.
Formation and rheological behavior of wormlike micelles in a catanionic system of fluoroacetic acid and tetradecyldimethylaminoxide.
Soft matter (2016-12-24)
Juan Zhang, Yingxia Wang, Guiying Xu, Meinqin Lin, Tianyu Fan, Zihao Yang, Zhaoxia Dong
ABSTRACT
From catanionic fluoro-/hydro-carbon mixtures of fluoroacetic acid (CF
Sigma-Aldrich
Fluoroacetic acid, 95%