Merck
  • Naloxegol, an opioid antagonist with reduced CNS penetration: Mode-of-action and human relevance for rat testicular tumours.
Naloxegol, an opioid antagonist with reduced CNS penetration: Mode-of-action and human relevance for rat testicular tumours.
Toxicology and applied pharmacology (2017-05-31)
Håkan Andersson, Terri Mitchard, Nakpangi Johnson, Eike Floettmann
ABSTRACT
Naloxegol is an opioid antagonist which has been developed for the treatment of patients with opioid induced constipation. In the nonclinical safety program naloxegol was shown to have a very benign toxicity profile. In the rat, but not the mouse, 2-year carcinogenicity study a change in tumour pattern with an increase in testicular Leydig cell tumours (LCT) was observed after dosing at high (supra-pharmacological) concentrations. To establish the basis of the increase in LCT and to assess its potential relevance to humans, studies to exclude and potentially identify mode-of-action (MoA) were performed. A genotoxic mechanism was ruled out following negative results in the Ames, mouse lymphoma, and micronucleus assays. An effect on androgen metabolism was excluded since the treatment of rats with naloxegol for 14days did not result in any induction of CYP protein levels. It was demonstrated that administration of centrally restricted opioid antagonists naloxegol or methylnaltrexone at high doses induced an increase in LH release with no clear increase in testosterone, in contrast to the centrally acting opioid antagonist naloxone, which showed marked increases in both LH and testosterone. LCT due to increased LH stimulation is common in rats but not documented in humans. Collectively, the lack of genotoxicity signal, the lack of androgen effect, the increase in LH secretion in rats, which is no considered to be relevant for LCT formation in humans, and high margins to clinical exposures, the observed increase in LCT in the rat is not expected to be clinically relevant.
Sigma-Aldrich
Human LHB / Lutropin Subunit Beta ELISA Kit