Merck
  • Role for reactive oxygen species in flow-stimulated inner medullary collecting duct endothelin-1 production.
Role for reactive oxygen species in flow-stimulated inner medullary collecting duct endothelin-1 production.
American journal of physiology. Renal physiology (2017-05-19)
Will Wheatley, Donald E Kohan
ABSTRACT
Inner medullary collecting duct (IMCD)-derived endothelin-1 (ET-1) is stimulated by volume expansion, in part through augmented luminal flow, whereupon it can elicit natriuresis and diuresis. Since flow can alter nitric oxide (NO) and reactive oxygen species (ROS), both of which can affect collecting duct salt transport, we asked whether NO and/or ROS mediate flow-stimulated IMCD ET-1. Mouse IMCD3 cells were exposed to flow, and ET-1/GAPDH mRNA was assessed. A shear stress of 10 dyn/cm
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder