Merck
  • Poly(ADP-ribose) polymerases regulate cell division and development in Arabidopsis roots.
Poly(ADP-ribose) polymerases regulate cell division and development in Arabidopsis roots.
Journal of integrative plant biology (2017-03-07)
Caifeng Liu, Qiao Wu, Weiwei Liu, Zongyin Gu, Wenjing Wang, Ping Xu, Hong Ma, Xiaochun Ge
ABSTRACT
Root organogenesis involves cell division, differentiation and expansion. The molecular mechanisms regulating root development are not fully understood. In this study, we identified poly(adenosine diphosphate (ADP)-ribose) polymerases (PARPs) as new players in root development. PARP catalyzes poly(ADP-ribosyl)ation of proteins by repeatedly adding ADP-ribose units onto proteins using nicotinamide adenine dinucleotide (NAD
Sigma-Aldrich
PARP1 Active human, recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)