Merck
  • Reduced expression of adipose triglyceride lipase decreases arachidonic acid release and prostacyclin secretion in human aortic endothelial cells.
Reduced expression of adipose triglyceride lipase decreases arachidonic acid release and prostacyclin secretion in human aortic endothelial cells.
Archives of physiology and biochemistry (2017-04-04)
Monika Riederer, Margarete Lechleitner, Harald Köfeler, Saša Frank
ABSTRACT
Vascular endothelial cells represent an important source of arachidonic acid (AA)-derived mediators involved in the generation of anti- or proatherogenic environments. Evidence emerged (in mast cells), that in addition to phospholipases, neutral lipid hydrolases as adipose triglyceride lipase (ATGL) also participate in this process. To examine the impact of ATGL on AA-release from cellular phospholipids (PL) and on prostacyclin secretion in human aortic endothelial cells (HAEC). siRNA-mediated silencing of ATGL promoted lipid droplet formation and TG accumulation in HAEC (nile red stain). ATGL knockdown decreased the basal and A23187 (calcium ionophore)-induced release of In vascular endothelial cells, the efficiency of stimulus-induced AA release and prostacyclin secretion is dependent on ATGL.
Sigma-Aldrich
MISSION® esiRNA, targeting human PNPLA2