Merck
  • Potential malignant transformation in the gastric mucosa of immunodeficient mice with persistent Mycoplasma penetrans infection.
Potential malignant transformation in the gastric mucosa of immunodeficient mice with persistent Mycoplasma penetrans infection.
PloS one (2017-07-12)
Shuyan Cao, Dandan Shen, Yadong Wang, Linxi Li, Liping Zhou, Yuxue Wang
ABSTRACT
Mycoplasma infection has been reported in immunocompromised cancer patients; nevertheless, it is not clear if persistent Mycoplasma infection could facilitate the proliferation of cancer cells in immunocompromised organisms. The aim of this study was to examine the relationship between persistent Mycoplasma infection and malignant transformation in an immunodeficient host model. Immunodeficient mouse model was established using cyclophosphamide and mice gastric mucosal cells were infected with Mycoplasma penetrans (Mpe). After 18 weeks, mice were sacrificed and gastric mucosal Mpe infected cells were identified by fluorescence in situ hybridization (FISH). Moreover, pathological and ultrastructural changes in mice gastric mucosa were evaluated and the expression of multiple proto-oncogenes was examined by Western blot. Our data show that Mpe infection was detected in the blood of immunodeficient mice and Mpe persistent infection in mice gastric mucosa was confirmed by FISH. There were pathological and ultrastructural malignant transformation occurred in the gastric mucosa of infected mice compared to control mice. Mpe infected mice showed lower expression of p53 and p21 and higher H-ras expression compared to the control group. Moreover, expression of NF-κB p65 subunit increased in Mpe infected mice, similar to the TNF-α expression. Bax expression in gastric mucosa of Mpe infected mice was lower while Bcl-2 expression was higher than in the uninfected control group. Collectively these data demonstrate that persistent Mpe infection is associated with aberrant expression of multiple proto-oncogenes in gastric mucosa of immunodeficient mice which potentially facilitate the malignant transformation.
Product Description
ECL Blotting Reagents, Cytiva RPN2109