Merck
  • Clinical value of R-spondins in triple-negative and metaplastic breast cancers.
Clinical value of R-spondins in triple-negative and metaplastic breast cancers.
British journal of cancer (2017-05-05)
F Coussy, F Lallemand, S Vacher, A Schnitzler, W Chemlali, M Caly, A Nicolas, S Richon, D Meseure, R El Botty, L De-Plater, L Fuhrmann, T Dubois, S Roman-Roman, V Dangles-Marie, E Marangoni, I Bièche
ABSTRACT
RSPO ligands, activators of the Wnt/β-catenin pathway, are overexpressed in different cancers. The objective of this study was to investigate the role of RSPOs in breast cancer (BC). Expression of RSPO and markers of various cancer pathways were measured in breast tumours and cell lines by qRT-PCR. The effect of RSPO on the Wnt/β-catenin pathway activity was determined by luciferase assay, western blotting, and qRT-PCR. The effect of RSPO2 inhibition on proliferation was determined by using RSPO2 siRNAs. The effect of IWR-1, an inhibitor of the Wnt/β-catenin pathway, was examined on the growth of an RSPO2-positive patient-derived xenograft (PDX) model of metaplastic triple-negative BC. We detected RSPO2 and RSPO4 overexpression levels in BC, particularly in triple-negative BC (TNBC), metaplastic BC, and triple-negative cell lines. Various mechanisms could account for this overexpression: presence of fusion transcripts involving RSPO, and amplification or hypomethylation of RSPO genes. Patients with RSPO2-overexpressing tumours have a poorer metastasis-free survival (P=3.6 × 10-4). RSPO2 and RSPO4 stimulate Wnt/β-catenin pathway activity. Inhibition of RSPO expression in a TN cell line inhibits cell growth, and IWR-1 significantly inhibits the growth of an RSPO2-overexpressing PDX. RSPO overexpression could therefore be a new prognostic biomarker and therapeutic target for TNBC.
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting human RSPO2