Merck
  • Highly efficient Blue-Emitting CdSe-derived Core/Shell Gradient Alloy Quantum Dots with Improved Photoluminescent Quantum Yield and Enhanced Photostability.
Highly efficient Blue-Emitting CdSe-derived Core/Shell Gradient Alloy Quantum Dots with Improved Photoluminescent Quantum Yield and Enhanced Photostability.
Langmuir : the ACS journal of surfaces and colloids (2017-04-01)
Junsang Cho, Yun Ku Jung, Jin-Kyu Lee, Hak-Sung Jung
ABSTRACT
Highly efficient blue-emitting CdSe-derived core/shell gradient alloy quantum dots (CSGA QDs) with photoluminescence quantum yield (PL QY) of ca. 90% have been synthesized through a facile "one-pot" approach. CdSe nuclei are initially formed as core and gradient alloy shells such as CdSe
Sigma-Aldrich
Zinc acetate, 99.99% trace metals basis