- [Mn(CO)4{S2CNMe(CH2CO2H)}], a new water-soluble CO-releasing molecule.
[Mn(CO)4{S2CNMe(CH2CO2H)}], a new water-soluble CO-releasing molecule.
Dalton transactions (Cambridge, England : 2003) (2011-03-16)
Sian H Crook, Brian E Mann, Anthony J H M Meijer, Harry Adams, Philip Sawle, David Scapens, Roberto Motterlini
PMID21403944
ABSTRACT
[Mn(CO)(4){S(2)CNMe(CH(2)CO(2)H)}], 1, is shown to be a CO releasing molecule providing at least three moles CO per mole of compound. The mechanism of CO loss is dissociative and reversible and was investigated using Gaussian 09 calculations. The reversible binding of CO results in a relatively stable solution of the compound, while in the presence of a CO receptor or a ligand to prevent the rebinding of CO, the CO is lost rapidly. The X-ray structure was determined.
MATERIALS