- CYP450 Enzymes Effect Oxygen-Dependent Reduction of Azide-Based Fluorogenic Dyes.
CYP450 Enzymes Effect Oxygen-Dependent Reduction of Azide-Based Fluorogenic Dyes.
ACS central science (2017-02-06)
Liam J O'Connor, Ishna N Mistry, Sarah L Collins, Lisa K Folkes, Graham Brown, Stuart J Conway, Ester M Hammond
PMID28149949
ABSTRACT
Azide-containing compounds have broad utility in organic synthesis and chemical biology. Their use as powerful tools for the labeling of biological systems
MATERIALS