Merck
  • Synthesis, characterization and biological activity of Cu(II), Ni(II) and Zn(II) complexes of biopolymeric Schiff bases of salicylaldehydes and chitosan.
Synthesis, characterization and biological activity of Cu(II), Ni(II) and Zn(II) complexes of biopolymeric Schiff bases of salicylaldehydes and chitosan.
International journal of biological macromolecules (2016-11-17)
Eliene Leandro de Araújo, Hellen Franciane Gonçalves Barbosa, Edward Ralph Dockal, Éder Tadeu Gomes Cavalheiro
ABSTRACT
Schiff bases have been prepared from biopolymer chitosan and salicylaldehyde, 5-methoxysalicylaldehyde, and 5-nitrosalicylaldehyde. Ligands were synthesized in a 1:1.5mol ratio, and their Cu(II), Ni(II) and Zn(II) complexes in a 1:1mol ratio (ligand:metal). Ligands were characterized by
Sigma-Aldrich
2-Hydroxy-5-methoxybenzaldehyde, 98%
Sigma-Aldrich
2-Hydroxy-5-nitrobenzaldehyde, 98%