- Instant detection and identification of concealed explosive-related compounds: Induced Stokes Raman versus infrared.
Instant detection and identification of concealed explosive-related compounds: Induced Stokes Raman versus infrared.
Forensic science international (2016-12-07)
Sherif Elbasuney, Ashraf F El-Sherif
PMID27923170
ABSTRACT
The instant detection of explosives and explosive-related compounds has become an urgent priority in recent years for homeland security and counter-terrorism applications. Modern techniques should offer enhancement in selectivity, sensitivity, and standoff distances. Miniaturisation, portability, and field-ruggedisation are crucial requirements. This study reports on instant and standoff identification of concealed explosive-related compounds using customized Raman technique. Stokes Raman spectra of common explosive-related compounds were generated and spectrally resolved to create characteristic finger print spectra. The scattered Raman emissions over the band 400:2000cm
MATERIALS