Merck
  • Daxx inhibits hypoxia-induced lung cancer cell metastasis by suppressing the HIF-1α/HDAC1/Slug axis.
Daxx inhibits hypoxia-induced lung cancer cell metastasis by suppressing the HIF-1α/HDAC1/Slug axis.
Nature communications (2016-12-23)
Ching-Wen Lin, Lu-Kai Wang, Shu-Ping Wang, Yi-Liang Chang, Yi-Ying Wu, Hsuan-Yu Chen, Tzu-Hung Hsiao, Wei-Yun Lai, Hsuan-Hsuan Lu, Ya-Hsuan Chang, Shuenn-Chen Yang, Ming-Wei Lin, Chi-Yuan Chen, Tse-Ming Hong, Pan-Chyr Yang
ABSTRACT
Hypoxia is a major driving force of cancer invasion and metastasis. Here we show that death domain-associated protein (Daxx) acts to negatively regulate hypoxia-induced cell dissemination and invasion by inhibiting the HIF-1α/HDAC1/Slug pathway. Daxx directly binds to the DNA-binding domain of Slug, impeding histone deacetylase 1 (HDAC1) recruitment and antagonizing Slug E-box binding. This, in turn, stimulates E-cadherin and occludin expression and suppresses Slug-mediated epithelial-mesenchymal transition (EMT) and cell invasiveness. Under hypoxic conditions, stabilized hypoxia-inducible factor (HIF)-1α downregulates Daxx expression and promotes cancer invasion, whereas re-expression of Daxx represses hypoxia-induced cancer invasion. Daxx also suppresses Slug-mediated lung cancer metastasis in an orthotopic lung metastasis mouse model. Using clinical tumour samples, we confirmed that the HIF-1α/Daxx/Slug pathway is an outcome predictor. Our results support that Daxx can act as a repressor in controlling HIF-1α/HDAC1/Slug-mediated cancer cell invasion and is a potential therapeutic target for inhibition of cancer metastasis.
Sigma-Aldrich
ChIPAb+ HDAC1 Antibody, rabbit polyclonal, from rabbit
Sigma-Aldrich
Anti-DAXX antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.