Merck
  • Characterization of Integrons and qnr Genes in Proteeae from a Teaching Hospital in China.
Characterization of Integrons and qnr Genes in Proteeae from a Teaching Hospital in China.
Chemotherapy (2016-05-26)
Jianming Cao, Meimei Li, Chunquan Xu, Tieli Zhou, Jia Du, Yao Sun, Ling Qin, Jiru Xu
ABSTRACT
Proteeae isolates displaying multidrug-resistance (MDR) are the second most common causes of hospital-associated infections. The aim of this study was to screen class 1-3 integrons and plasmid-mediated quinolone resistance (PMQR) genes in Proteeae isolates from the First Affiliated Hospital of the Wenzhou Medical University. 176 Proteeae isolates were collected from clinical specimens of inpatients between January 2011 and December 2013. Susceptibility testing was determined by the agar dilution method. Class 1-3 integrons and PMQR genes were amplified by polymerase chain reaction, and the variable regions of integrons were determined by restriction fragment length polymorphisms. 68.2% Proteeae isolates exhibited MDR phenotypes: 46.6 and 10.8% Proteeae isolates were positive for intI1 and intI2, respectively. The resistance rate of integron-positive isolates to aminoglycosides, fluoroquinolones, and trimethoprim/sulfamethoxazole was significantly higher than integron-negative isolates. Sequence analysis revealed that dfrA1-sat2-aadA1, dfrA1-catB2-sat2-aadA1, and sat2-aadA1 were first detected in Morganella morganii strains isolated from China. PMQR was determined by qnrD in 40 strains (22.7%). Our results indicate that class 1 and 2 integrons are common among Proteeae isolates. Meanwhile, qnrD are highly prevalent in Proteeae isolated from our hospital.