Merck
  • Transcription Factor ZBP-89 Drives a Feedforward Loop of β-Catenin Expression in Colorectal Cancer.
Transcription Factor ZBP-89 Drives a Feedforward Loop of β-Catenin Expression in Colorectal Cancer.
Cancer research (2016-10-21)
Bryan E Essien, Sinju Sundaresan, Ramon Ocadiz-Ruiz, Aaron Chavis, Amy C Tsao, Arthur J Tessier, Michael M Hayes, Amanda Photenhauer, Milena Saqui-Salces, Anthony J Kang, Yatrik M Shah, Balazs Győrffy, Juanita L Merchant
ABSTRACT
In colorectal cancer, APC-mediated induction of unregulated cell growth involves posttranslational mechanisms that prevent proteasomal degradation of proto-oncogene β-catenin (CTNNB1) and its eventual translocation to the nucleus. However, about 10% of colorectal tumors also exhibit increased CTNNB1 mRNA. Here, we show in colorectal cancer that increased expression of ZNF148, the gene coding for transcription factor ZBP-89, correlated with reduced patient survival. Tissue arrays showed that ZBP-89 protein was overexpressed in the early stages of colorectal cancer. Conditional deletion of Zfp148 in a mouse model of Apc-mediated intestinal polyps demonstrated that ZBP-89 was required for polyp formation due to induction of Ctnnb1 gene expression. Chromatin immunoprecipitation (ChIP) and EMSA identified a ZBP-89-binding site in the proximal promoter of CTNNB1 Reciprocally, siRNA-mediated reduction of CTNNB1 expression also decreased ZBP-89 protein. ChIP identified TCF DNA binding sites in the ZNF148 promoter through which Wnt signaling regulates ZNF148 gene expression. Suppression of either ZNF148 or CTNNB1 reduced colony formation in WNT-dependent, but not WNT-independent cell lines. Therefore, the increase in intracellular β-catenin protein initiated by APC mutations is sustained by ZBP-89-mediated feedforward induction of CTNNB1 mRNA. Cancer Res; 76(23); 6877-87. ©2016 AACR.
Sigma-Aldrich
MISSION® esiRNA, targeting human CTNNB1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
EZ-Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads. Control primers included.