Merck
  • Comparative analysis of viral RNA signatures on different RIG-I-like receptors.
Comparative analysis of viral RNA signatures on different RIG-I-like receptors.
eLife (2016-03-25)
Raul Y Sanchez David, Chantal Combredet, Odile Sismeiro, Marie-Agnès Dillies, Bernd Jagla, Jean-Yves Coppée, Marie Mura, Mathilde Guerbois Galla, Philippe Despres, Frédéric Tangy, Anastassia V Komarova
ABSTRACT
The RIG-I-like receptors (RLRs) play a major role in sensing RNA virus infection to initiate and modulate antiviral immunity. They interact with particular viral RNAs, most of them being still unknown. To decipher the viral RNA signature on RLRs during viral infection, we tagged RLRs (RIG-I, MDA5, LGP2) and applied tagged protein affinity purification followed by next-generation sequencing (NGS) of associated RNA molecules. Two viruses with negative- and positive-sense RNA genome were used: measles (MV) and chikungunya (CHIKV). NGS analysis revealed that distinct regions of MV genome were specifically recognized by distinct RLRs: RIG-I recognized defective interfering genomes, whereas MDA5 and LGP2 specifically bound MV nucleoprotein-coding region. During CHIKV infection, RIG-I associated specifically to the 3' untranslated region of viral genome. This study provides the first comparative view of the viral RNA ligands for RIG-I, MDA5 and LGP2 in the presence of infection.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
TRI Reagent®, LS, For processing fluid samples such as cell suspensions, CSF, and amniotic fluid.
Pricing and availability is not currently available.