Merck
  • Restoration of Hydrogen Sulfide Production in Diabetic Mice Improves Reparative Function of Bone Marrow Cells.
Restoration of Hydrogen Sulfide Production in Diabetic Mice Improves Reparative Function of Bone Marrow Cells.
Circulation (2016-09-24)
Zhongjian Cheng, Venkata Naga Srikanth Garikipati, Emily Nickoloff, Chunlin Wang, David J Polhemus, Jibin Zhou, Cynthia Benedict, Mohsin Khan, Suresh K Verma, Joseph E Rabinowitz, David Lefer, Raj Kishore
ABSTRACT
Bone marrow cell (BMC)-based treatment for critical limb ischemia in diabetic patients yielded a modest therapeutic effect resulting from cell dysfunction. Therefore, approaches that improve diabetic stem/progenitor cell functions may provide therapeutic benefits. Here, we tested the hypothesis that restoration of hydrogen sulfide (H Mouse BMCs were isolated by density-gradient centrifugation. Unilateral hind limb ischemia was conducted in 12- to 14-week-old db/+ and db/db mice by ligation of the left femoral artery. The H Both H Decreased CSE-mediated H
Sigma-Aldrich
MISSION® esiRNA, targeting human CTH