Merck
  • Synthesis and evaluation of copper(II) complexes with isoniazid-derived hydrazones as anticancer and antitubercular agents.
Synthesis and evaluation of copper(II) complexes with isoniazid-derived hydrazones as anticancer and antitubercular agents.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2016-09-07)
Gisele S S Firmino, Marcus V N de Souza, Claudia Pessoa, Maria C S Lourenco, Jackson A L C Resende, Josane A Lessa
ABSTRACT
In this study, the N,N,O metal chelator 2-pyridinecarboxaldehydeisonicotinoyl hydrazone (HPCIH, 1) and its derivatives 2-acetylpyridine-(HAPIH 2), 2-pyridineformamide-(HPAmIH, 3) and pyrazineformamide-(HPzAmIH, 4) were employed in the synthesis of four copper(II) complexes, [Cu(HPCIH)Cl
Sigma-Aldrich
Pyrazinecarbonitrile, 99%
Sigma-Aldrich
2-Pyridinecarboxaldehyde, 99%