Merck
  • Impairment in subcortical suppression in schizophrenia: Evidence from the fBIRN Oddball Task.
Impairment in subcortical suppression in schizophrenia: Evidence from the fBIRN Oddball Task.
Human brain mapping (2016-08-02)
Katie M Lavigne, Mahesh Menon, Todd S Woodward
ABSTRACT
Schizophrenia patients show widespread impairments in brain activity during oddball tasks, which involve responding to infrequent target stimuli while refraining from responding during continuous non-target stimuli. In a network-based investigation comparing schizophrenia or schizoaffective patients to healthy controls, we sought to clarify which networks were specifically associated with target detection using a multivariate analysis technique that identifies task-specific functional brain networks. We acquired data from the publicly available function biomedical informatics research network collaboration, including 58 patients and 50 controls. Two task-based functional brain networks were identified: (1) a response modulation network including bilateral temporal pole, supramarginal gyrus, striatum, and thalamus, on which patients showed decreased activity relative to controls; and (2) an auditory-motor response activation network, on which patients showed a slower return to baseline than controls, but no difference in peak activation. For both groups, baseline to peak activation of the response modulation network correlated negatively with peak to baseline activity in the response activation network, suggesting a role in suppressing the motor response following targets. Patients' impaired activity in the response modulation network, and subsequent longer return to baseline in the response activation network, correspond with their later and less accurate behavioral performance, suggesting that impairment in suppression of the auditory-motor response activation network could underlie oddball task deficits in schizophrenia. In addition, the magnitude of the activity in the response modulation network was correlated with intensity of delusions of reference, supporting the notion that increased referential ideation is associated with hyperactivity within the subcortical striatal-limbic network. Hum Brain Mapp 37:4640-4653, 2016. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
2,2′:5′,2′′:5′′,2′′′-Quaterthiophene, 96%