Merck
  • Effects of nicotine on bone during orthodontic tooth movement in male rats. Histological and immunohistochemical study.
Effects of nicotine on bone during orthodontic tooth movement in male rats. Histological and immunohistochemical study.
Saudi medical journal (2016-09-22)
Manal A Bakathir, Amal I Linjawi, Samia S Omar, Ahad B Aboqura, Ali H Hassan
ABSTRACT
To investigate the effects of nicotine on orthodontic tooth movement and accompanying histological and immunohistochemical changes in rats. An experimental study conducted at King Abdulaziz University, Jeddah, Saudi Arabia between 2013 and 2014. Thirty-two rats randomly divided into 4 groups. Three were experimental, received daily nicotine injections: group A: 0.37 mg/kg, group B: 0.57 mg/kg, and group C: 0.93 mg/kg. The control group  (group D) received a daily injection of 0.5 mL saline. All rats were subjected to 30 g of orthodontic force on the maxillary left first molars and incisors using a nickel-titanium closed-coil spring. The distance between the 2 teeth was assessed before and after 14 days of force application. Histological, immunohistochemical, and histomorphometric assessments were performed on sections from groups C and D. Groups C (p less than 0.001) and D (p less than 0.001) showed the significantly greatest and least amounts of tooth movement . The results were statistically dose-dependent. Unbalanced resorption-apposition bone remodeling patterns and increased osteoclast cell distribution were observed in the nicotine group with significantly smaller percentages of bone surface areas mesially and distally (p less than 0.05). Immunohistochemical stains showed low alkaline phosphatase activity and intense tartrate-resistant acid phosphatase activity in the nicotine group. Nicotine accelerated orthodontic tooth movement with unbalanced bone resorption and apposition patterns around the moving teeth.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Nicotine ditartrate, European Pharmacopoeia (EP) Reference Standard