Merck
  • CD16 is indispensable for antibody-dependent cellular cytotoxicity by human monocytes.
CD16 is indispensable for antibody-dependent cellular cytotoxicity by human monocytes.
Scientific reports (2016-09-28)
Wei Hseun Yeap, Kok Loon Wong, Noriko Shimasaki, Esmeralda Chi Yuan Teo, Jeffrey Kim Siang Quek, Hao Xiang Yong, Colin Phipps Diong, Antonio Bertoletti, Yeh Ching Linn, Siew Cheng Wong
ABSTRACT
Antibody-dependent cellular cytotoxicity (ADCC) is exerted by immune cells expressing surface Fcγ receptors (FcγRs) against cells coated with antibody, such as virus-infected or transformed cells. CD16, the FcγRIIIA, is essential for ADCC by NK cells, and is also expressed by a subset of human blood monocytes. We found that human CD16- expressing monocytes have a broad spectrum of ADCC capacities and can kill cancer cell lines, primary leukemic cells and hepatitis B virus-infected cells in the presence of specific antibodies. Engagement of CD16 on monocytes by antibody bound to target cells activated β2-integrins and induced TNFα secretion. In turn, this induced TNFR expression on the target cells, making them susceptible to TNFα-mediated cell death. Treatment with TLR agonists, DAMPs or cytokines, such as IFNγ, further enhanced ADCC. Monocytes lacking CD16 did not exert ADCC but acquired this property after CD16 expression was induced by either cytokine stimulation or transient transfection. Notably, CD16+ monocytes from patients with leukemia also exerted potent ADCC. Hence, CD16+ monocytes are important effectors of ADCC, suggesting further developments of this property in the context of cellular therapies for cancer and infectious diseases.
Sigma-Aldrich
Sodium chromate, 98%
Pricing and availability is not currently available.