- Optical Control of Peroxisomal Trafficking.
Optical Control of Peroxisomal Trafficking.
ACS synthetic biology (2015-10-30)
Jessica I Spiltoir, Devin Strickland, Michael Glotzer, Chandra L Tucker
PMID26513473
ABSTRACT
The blue-light-responsive LOV2 domain of Avena sativa phototropin1 (AsLOV2) has been used to regulate activity and binding of diverse protein targets with light. Here, we used AsLOV2 to photocage a peroxisomal targeting sequence, allowing light regulation of peroxisomal protein import. We generated a protein tag, LOV-PTS1, that can be appended to proteins of interest to direct their import to the peroxisome with light. This method provides a means to inducibly trigger peroxisomal protein trafficking in specific cells at user-defined times.
Sigma-Aldrich
Anti-PMP70 antibody, Mouse monoclonal, clone 70-18, purified from hybridoma cell culture