Merck
  • Regulation of Cell Migration and β1 Integrin Trafficking by the Endosomal Adaptor GGA3.
Regulation of Cell Migration and β1 Integrin Trafficking by the Endosomal Adaptor GGA3.
Traffic (Copenhagen, Denmark) (2016-03-05)
Colin D H Ratcliffe, Pranshu Sahgal, Christine A Parachoniak, Johanna Ivaska, Morag Park
ABSTRACT
The integrin family of cell adhesion receptors link extracellular matrices to intracellular signaling pathways and the actin cytoskeleton; and regulate cell migration, proliferation and survival in normal and diseased tissues. The subcellular location of integrin receptors is critical for their function and deregulated trafficking is implicated in various human diseases. Here we identify a role for Golgi-localized gamma-ear containing Arf-binding protein 3 (GGA3), in regulating trafficking of β1 integrin. GGA3 knockdown reduces cell surface and total levels of α2, α5 and β1 integrin subunits, inhibits cell spreading, reduces focal adhesion number, as well as cell migration. In the absence of GGA3, integrins are increasingly retained inside the cell, traffic toward the perinuclear lysosomal compartment and their degradation is enhanced. Integrin traffic and maintenance of integrin levels are dependent on the integrity of the Arf binding site of GGA3. Furthermore, sorting nexin 17 (SNX17), a critical regulator of integrin recycling, becomes mislocalized to enlarged late endosomes upon GGA3 depletion. These data support a model whereby GGA3, through its ability to regulate SNX17 endosomal localization and through interaction with Arf6 diverts integrins from the degradative pathway supporting cell migration.
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αv (Ab-1) Mouse mAb (272-17E6), liquid, clone 272-17E6, Calbiochem®