Merck
  • Hall effect in the accumulation layers on the surface of organic semiconductors.
Hall effect in the accumulation layers on the surface of organic semiconductors.
Physical review letters (2005-12-31)
V Podzorov, E Menard, J A Rogers, M E Gershenson
ABSTRACT
We have observed the Hall effect in the field-induced accumulation layer on the surface of single-crystal samples of a small-molecule organic semiconductor rubrene. The Hall mobility muH increases with decreasing temperature in both the intrinsic (high-temperature) and trap-dominated (low-temperature) conduction regimes. In the intrinsic regime, the density of mobile field-induced charge carriers extracted from the Hall measurements, nH, coincides with the density n calculated using the gate-channel capacitance and becomes smaller than n in the trap-dominated regime. The Hall data are consistent with the diffusive bandlike motion of field-induced charge carriers between trapping events.
Sigma-Aldrich
Rubrene, sublimed grade, 99.99% trace metals basis