Merck
  • The enantioselective organocatalytic 1,4-addition of electron-rich benzenes to alpha,beta-unsaturated aldehydes.
The enantioselective organocatalytic 1,4-addition of electron-rich benzenes to alpha,beta-unsaturated aldehydes.
Journal of the American Chemical Society (2002-07-04)
Nick A Paras, David W C MacMillan
ABSTRACT
The first enantioselective organocatalytic alkylation of electron-rich benzene rings with alpha,beta-unsaturated aldehydes has been accomplished. The use of iminium catalysis has provided a new strategy for the enantioselective construction of benzylic stereogenicity, an important chiral synthon for natural product and medicinal agent synthesis. The (2S,5S)-5-benzyl-2-tert-butylimidazolidinone amine catalyst has been found to mediate the conjugate addition of a wide variety of substituted and unsubstituted anilines to unsaturated aldehydes. A diverse spectrum of aldehyde substrates can also be accommodated in this new organocatalytic transformation. While catalyst quantities of 10 mol % were generally employed in this study, successful alkylations conducted with catalyst loadings as low as 1 mol % are described.
Sigma-Aldrich
(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone, 97%