- Formation of nickeladihydropyran by oxidative addition of cyclopropyl ketone. Key intermediate in nickel-catalyzed cycloaddition.
Formation of nickeladihydropyran by oxidative addition of cyclopropyl ketone. Key intermediate in nickel-catalyzed cycloaddition.
Journal of the American Chemical Society (2006-04-20)
Sensuke Ogoshi, Midue Nagata, Hideo Kurosawa
PMID16620100
ABSTRACT
Cyclopropyl phenyl ketone underwent oxidative addition to Ni(PCy3) generated from Ni(cod)2 and PCy3 to give a nickeladihydropyran, which is a key intermediate for the Ni(0)-catalyzed homo- or heterocycloaddition to give cyclopentane compounds having two carbonyl substituents at the 1,3-position.
MATERIALS