Merck
  • Mimicking the germinal center reaction in hybridoma cells to isolate temperature-selective anti-PEG antibodies.
Mimicking the germinal center reaction in hybridoma cells to isolate temperature-selective anti-PEG antibodies.
mAbs (2014-05-31)
Yu-Cheng Su, Talal S Al-Qaisi, Hsin-Yi Tung, Tian-Lu Cheng, Kuo-Hsiang Chuang, Bing-Mae Chen, Steve R Roffler
ABSTRACT
Modification of antibody class and binding properties typically requires cloning of antibody genes, antibody library construction, phage or yeast display and recombinant antibody expression. Here, we describe an alternative "cloning-free" approach to generate antibodies with altered antigen-binding and heavy chain isotype by mimicking the germinal center reaction in antibody-secreting hybridoma cells. This was accomplished by lentiviral transduction and controllable expression of activation-induced cytidine deaminase (AID) to generate somatic hypermutation and class switch recombination in antibody genes coupled with high-throughput fluorescence-activated cell sorting (FACS) of hybridoma cells to detect altered antibody binding properties. Starting from a single established hybridoma clone, we isolated mutated antibodies that bind to a low-temperature structure of polyethylene glycol (PEG), a polymer widely used in nanotechnology, biotechnology and pharmaceuticals. FACS of AID-infected hybridoma cells also facilitated rapid identification of class switched variants of monoclonal IgM to monoclonal IgG. Mimicking the germinal center reaction in hybridoma cells may offer a general method to identify and isolate antibodies with altered binding properties and class-switched heavy chains without the need to carry out DNA library construction, antibody engineering and recombinant protein expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human TM4SF1