Merck
  • Hepatocyte-targeted RNAi therapeutics for the treatment of chronic hepatitis B virus infection.
Hepatocyte-targeted RNAi therapeutics for the treatment of chronic hepatitis B virus infection.
Molecular therapy : the journal of the American Society of Gene Therapy (2013-02-27)
Christine I Wooddell, David B Rozema, Markus Hossbach, Matthias John, Holly L Hamilton, Qili Chu, Julia O Hegge, Jason J Klein, Darren H Wakefield, Claudia E Oropeza, Jochen Deckert, Ingo Roehl, Kerstin Jahn-Hofmann, Philipp Hadwiger, Hans-Peter Vornlocher, Alan McLachlan, David L Lewis
ABSTRACT
RNA interference (RNAi)-based therapeutics have the potential to treat chronic hepatitis B virus (HBV) infection in a fundamentally different manner than current therapies. Using RNAi, it is possible to knock down expression of viral RNAs including the pregenomic RNA from which the replicative intermediates are derived, thus reducing viral load, and the viral proteins that result in disease and impact the immune system's ability to eliminate the virus. We previously described the use of polymer-based Dynamic PolyConjugate (DPC) for the targeted delivery of siRNAs to hepatocytes. Here, we first show in proof-of-concept studies that simple coinjection of a hepatocyte-targeted, N-acetylgalactosamine-conjugated melittin-like peptide (NAG-MLP) with a liver-tropic cholesterol-conjugated siRNA (chol-siRNA) targeting coagulation factor VII (F7) results in efficient F7 knockdown in mice and nonhuman primates without changes in clinical chemistry or induction of cytokines. Using transient and transgenic mouse models of HBV infection, we show that a single coinjection of NAG-MLP with potent chol-siRNAs targeting conserved HBV sequences resulted in multilog repression of viral RNA, proteins, and viral DNA with long duration of effect. These results suggest that coinjection of NAG-MLP and chol-siHBVs holds great promise as a new therapeutic for patients chronically infected with HBV.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 25 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.