Merck
  • Cytoplasmic p21(Cip1/WAF1) regulates neurite remodeling by inhibiting Rho-kinase activity.
Cytoplasmic p21(Cip1/WAF1) regulates neurite remodeling by inhibiting Rho-kinase activity.
The Journal of cell biology (2002-07-18)
Hiroyuki Tanaka, Toshihide Yamashita, Minoru Asada, Shuki Mizutani, Hideki Yoshikawa, Masaya Tohyama
ABSTRACT
p21(Cip1/WAF1) has cell cycle inhibitory activity by binding to and inhibiting both cyclin/Cdk kinases and proliferating cell nuclear antigen. Here we show that p21(Cip1/WAF1) is induced in the cytoplasm during the course of differentiation of chick retinal precursor cells and N1E-115 cells. Ectopic expression of p21(Cip1/WAF1) lacking the nuclear localization signal in N1E-115 cells and NIH3T3 cells affects the formation of actin structures, characteristic of inactivation of Rho. p21(Cip1/WAF1) forms a complex with Rho-kinase and inhibits its activity in vitro and in vivo. Neurite outgrowth and branching from the hippocampal neurons are promoted if p21(Cip1/WAF1) is expressed abundantly in the cytoplasm. These results suggest that cytoplasmic p21(Cip1/WAF1) may contribute to the developmental process of the newborn neurons that extend axons and dendrites into target regions.
Sigma-Aldrich
ROKα/ROCK-II Protein, active, rat, 10 µg, Active, rat, N-terminal, His-tagged, fusion protein corresponding to amino acids 2-543 of rat ROKa/ROCK-II. For use in Kinase Assays.