Merck
  • A useful Pd-catalyzed Negishi coupling approach to benzylic sulfonamide derivatives.
A useful Pd-catalyzed Negishi coupling approach to benzylic sulfonamide derivatives.
Organic letters (2008-05-22)
Gang Zhou, Pauline Ting, Robert Aslanian, John J Piwinski
ABSTRACT
A mild catalytic system to access diversely functionalized benzylic sulfonamides has been developed. Palladium-catalyzed alpha-arylation by Negishi cross-coupling of sulfonamide-stabilized anions and a wide range of aryl iodides, bromides, and triflates constitutes a practical strategy for the synthesis of various benzylic sulfonamides.
Sigma-Aldrich
4-Bromo-2-methylanisole, 98%