Merck
  • Irisin, a unique non-inflammatory myokine in stimulating skeletal muscle metabolism.
Irisin, a unique non-inflammatory myokine in stimulating skeletal muscle metabolism.
Journal of physiology and biochemistry (2015-09-25)
Roger A Vaughan, Nicholas P Gannon, Christine M Mermier, Carole A Conn
ABSTRACT
Exercise offers several benefits for health, including increased lean body mass and heightened energy expenditure, which may be partially attributable to secretory factors known as myokines. Irisin, a recently identified myokine, was shown to increase metabolic rate and mitochondrial content in both myocytes and adipocytes; however, the mechanism(s) of action still remain largely unexplained. This work investigated if irisin functions by acting as an inflammatory myokine leading to cellular stress and energy expenditure. C2C12 myotubes were treated with various concentrations of irisin, TNFα, or IL6 for various durations. Glycolytic and oxidative metabolism, as well as mitochondrial uncoupling, were quantified by measurement of acidification and oxygen consumption, respectively. Metabolic gene and protein expression were measured by quantitative real-time polymerase chain reaction (qRT-PCR) and immunoblotting, respectively. Mitochondrial content was assessed by fluorescent imaging. NFκB activity was assessed using an NFκB GFP-linked reporter system. Consistent with previous findings, irisin significantly increased expression of several genes including peroxisome proliferator-activated receptor γ coactivator-1α (PGC-1α) leading to increased mitochondrial content and oxygen consumption. Despite some similarities between TNFα and irisin treatment, irisin failed to activate the NFκB pathway like TNFα, suggesting that irisin may not act as an inflammatory signal. Irisin has several effects on myotube metabolism which appear to be dependent on substrate availability; however, the precise mechanism(s) by which irisin functions in skeletal muscle remain unclear. Our observations support the hypothesis that irisin does not function through inflammatory NFκB activation like other myokines (such as TNFα).
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
SAFC
Tromethamine
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
IL-6 from rat, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), ≥90% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis