Merck
  • shRNA Depletion of cIAP1 Sensitizes Human Ovarian Cancer Cells to Anticancer Agent-Induced Apoptosis.
shRNA Depletion of cIAP1 Sensitizes Human Ovarian Cancer Cells to Anticancer Agent-Induced Apoptosis.
Oncology research (2015-07-15)
Hong Jin, You-Yuan Dong, Hong Zhang, Ying Cui, Kai Xie, Ge Lou
ABSTRACT
Emerging evidence suggests a potential role of cellular inhibitor of apoptosis protein 1 (cIAP1) in the development of human ovarian cancer. However, its function in the progression of ovarian cancer has not been clearly determined. Our study aimed to investigate the effect of cIAP1 gene depletion on the chemosensitivity of ovarian cancer cells. We developed a novel short hairpin RNA (shRNA) plasmid specifically targeting cIAP1. Cell proliferation, invasion, and apoptosis of the shRNA-transfected cells were evaluated using MTT, Transwell chamber, and flow cytometric assays, respectively. The concentration of MMP-9 in the supernatant was detected by ELISA. Targeted depletion of cIAP1 by shRNA significantly reduced expression levels of cIAP1 mRNA and protein, leading to inhibition of cell proliferation and invasion capability in SKOV3 cells. At the same time, cIAP1 downregulation decreased the secretion of MMP-9. shRNA depletion of cIAP1 enhanced chemosensitivity of ovarian cancer cells to Taxol and carboplatin-induced apoptosis. cIAP1 is associated with tumor progression in human ovarian cancer. Therefore, cIAP1 might be a potential target for therapeutic anticancer drugs.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human BIRC2
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Carboplatin
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
MISSION® esiRNA, targeting human BIRC3
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Birc2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Birc3