- The crystal structure of hepatitis C virus NS3 proteinase reveals a trypsin-like fold and a structural zinc binding site.
The crystal structure of hepatitis C virus NS3 proteinase reveals a trypsin-like fold and a structural zinc binding site.
Cell (1996-10-18)
R A Love, H E Parge, J A Wickersham, Z Hostomsky, N Habuka, E W Moomaw, T Adachi, Z Hostomska
PMID8861916
ABSTRACT
During replication of hepatitis C virus (HCV), the final steps of polyprotein processing are performed by a viral proteinase located in the N-terminal one-third of nonstructural protein 3. The structure of NS3 proteinase from HCV BK strain was determined by X-ray crystallography at 2.4 angstrom resolution. NS3P folds as a trypsin-like proteinase with two beta barrels and a catalytic triad of His-57, Asp-81, Ser-139. The structure has a substrate-binding site consistent with the cleavage specificity of the enzyme. Novel features include a structural zinc-binding site and a long N-terminus that interacts with neighboring molecules by binding to a hydrophobic surface patch.