Merck
  • Design and solid phase synthesis of new DOTA conjugated (+)-biotin dimers planned to develop molecular weight-tuned avidin oligomers.
Design and solid phase synthesis of new DOTA conjugated (+)-biotin dimers planned to develop molecular weight-tuned avidin oligomers.
Organic & biomolecular chemistry (2015-02-28)
Alessandro Pratesi, Mauro Ginanneschi, Fabrizio Melani, Marco Chinol, Angela Carollo, Giovanni Paganelli, Marco Lumini, Mattia Bartoli, Marco Frediani, Luca Rosi, Giorgio Petrucci, Luigi Messori, Anna Maria Papini
ABSTRACT
Chemical modifications of the biotin carrier in pretargeted avidin–biotin radionuclide therapy may be of paramount importance for tuning the amount of the radioactivity delivered to cancer cells by labelled biotins. We report here the synthesis of a collection of new synthetic DOTA-constructs bearing two (+)-biotin molecules (bis-biotins), designed for the creation of multimeric Av units (tetramers) bonded to the antibody. All the syntheses were carried out following the solid phase strategy and growing the molecules on a Rink Amide resin. The biotin heads are connected through spacers containing PEG or non-PEG residues. Molecular modelling calculations suggested that the Av cross-linking ability of the bis-biotins depends mainly on the spacers length, with the best results being expected for arms affording distances in the range of 10–25 Å between the biotin carboxylate atoms, in the fully extended conformation. SEC-HPLC MALLS analysis of the products of our Av/bis-biotin reaction mixtures have confirmed this hypothesis. The bis-biotin 16, where the non-PEG linker ensured a distance of 26.7 Å between the biotin moieties, gave about 50% of Av oligomers while the shorter analogue 18 (19.5 Å) afforded 100% of an Av polymer containing about 21 protein units. Remarkably, the solubility of both the bis-biotins, i.e.16 and 18, in aqueous solutions was good and they showed excellent stability against the action of peptidases.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
Fmoc-Gly-OH, ≥98.0% (T)
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Supelco
Acetone, analytical standard
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%