Merck
Mapping pathological phenotypes in a mouse model of CDKL5 disorder.
PloS one (2014-05-20)
Elena Amendola, Yang Zhan, Camilla Mattucci, Enrico Castroflorio, Eleonora Calcagno, Claudia Fuchs, Giuseppina Lonetti, Davide Silingardi, Alexei L Vyssotski, Dominika Farley, Elisabetta Ciani, Tommaso Pizzorusso, Maurizio Giustetto, Cornelius T Gross
ABSTRACT
Mutations in cyclin-dependent kinase-like 5 (CDKL5) cause early-onset epileptic encephalopathy, a neurodevelopmental disorder with similarities to Rett Syndrome. Here we describe the physiological, molecular, and behavioral phenotyping of a Cdkl5 conditional knockout mouse model of CDKL5 disorder. Behavioral analysis of constitutive Cdkl5 knockout mice revealed key features of the human disorder, including limb clasping, hypoactivity, and abnormal eye tracking. Anatomical, physiological, and molecular analysis of the knockout uncovered potential pathological substrates of the disorder, including reduced dendritic arborization of cortical neurons, abnormal electroencephalograph (EEG) responses to convulsant treatment, decreased visual evoked responses (VEPs), and alterations in the Akt/rpS6 signaling pathway. Selective knockout of Cdkl5 in excitatory and inhibitory forebrain neurons allowed us to map the behavioral features of the disorder to separable cell-types. These findings identify physiological and molecular deficits in specific forebrain neuron populations as possible pathological substrates in CDKL5 disorder.
Sigma-Aldrich
Urethane, ≥99%
Supelco
Urethane, analytical standard
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard