Merck
  • Twenty-one-base-pair insertion polymorphism creates an enhancer element and potentiates SLC6A1 GABA transporter promoter activity.
Twenty-one-base-pair insertion polymorphism creates an enhancer element and potentiates SLC6A1 GABA transporter promoter activity.
Pharmacogenetics and genomics (2008-12-17)
Rungnapa Hirunsatit, Elizabeth D George, Barbara K Lipska, Hani M Elwafi, Lisa Sander, Carolyn M Yrigollen, Joel Gelernter, Elena L Grigorenko, Jaakko Lappalainen, Shrikant Mane, Angus C Nairn, Joel E Kleinman, Arthur A Simen
ABSTRACT
Sodium-dependent and chloride-dependent gamma-aminobutyric acid (GABA) transporter 1 (SLC6A1) is the target of a number of drugs of clinical importance and is a major determinant of synaptic GABA concentrations. We resequenced the human SLC6A1 gene previously and discovered a novel 21 bp insertion in the predicted promoter region that creates a second tandem copy of the sequence. Here we sought to determine the functional relevance of this variation. We used reporter assays, mobility shift assays, quantitative PCR, and proteomics methods as well as postmortem expression analysis for this work. Reporter assays showed that the insertion allele significantly increases promoter activity in multiple cell lines. The zinc finger transcription factor ZNF148 was found to significantly transactivate the promoter and increase expression when overexpressed but could not account for the differences in activity between the two alleles of the promoter. Copy number of the insertion sequence was associated with exponentially increasing activity of a downstream promoter, suggesting that the insertion sequence has enhancer activity when present in multiple copies. SLC6A1 promoter genotype was found to predict SLC6A1 RNA expression in human postmortem hippocampal samples. These results suggest that the insertion polymorphism leads to increased SLC6A1 promoter activity because, in part, of creation of an enhancer element when present as multiple copies. Genotyping individuals from Tanzania in this study suggested that the insertion allele has its origin in Africa. On account of the effect of the insertion on promoter activity, this relatively common polymorphism may prove useful in predicting clinical response to pharmacological modulators of SLC6A1 as well as GABAergic function in individuals of African descent.