Merck
  • Genetic variant in the 3'-untranslated region of VEGFR1 gene influences chronic obstructive pulmonary disease and lung cancer development in Chinese population.
Genetic variant in the 3'-untranslated region of VEGFR1 gene influences chronic obstructive pulmonary disease and lung cancer development in Chinese population.
Mutagenesis (2014-06-04)
Hui Wang, Lei Yang, Jieqiong Deng, Bo Wang, Xiaorong Yang, Rongrong Yang, Mei Cheng, Wenxiang Fang, Fuman Qiu, Xin Zhang, Weidong Ji, Pixin Ran, Yifeng Zhou, Jiachun Lu
ABSTRACT
Lung inflammation and epithelial to mesenchymal transition (EMT) are two pathogenic features for the two contextual diseases: chronic obstructive pulmonary disease (COPD) and lung cancer. VEGFR1 (or FLT1) plays a certain role in promoting tumour growth, inflammation and EMT. To simultaneously test the association between the single nucleotide polymorphisms (SNPs) in VEGFR1 and risk of COPD and lung cancer would reveal genetic mechanisms shared by these two diseases and joint aetiology. We conducted a two-population hospital-based case-control study. Three potential functional SNPs (rs664393, rs7326277 and rs9554314) were genotyped in southern Chinese and validated in eastern Chinese to explore their associations with COPD risk in 1511 COPD patients and 1677 normal lung function controls, and with lung cancer risk in 1559 lung cancer cases and 1679 cancer-free controls. We also detected the function of the promising SNP. Individuals carrying the rs7326277C (CT+CC) variant genotypes of VEGFR1 had a significant decrease in risk of both COPD (OR = 0.78; 95% CI = 0.68-0.90) and lung cancer (OR = 0.79; 95% CI = 0.64-0.98), compared with those carrying the rs7326277TT genotype. Functional assays further showed that the rs7326277C genotypes had lower transcriptional activity and caused decreased VEGFR expression, compared with the rs7326277TT genotype. However, no significant association was observed for the other two SNPs (rs664393 and rs9554314) and either COPD or lung cancer risk. Our data suggested that the rs7326277C variant of VEGFR1 could reduce both COPD and lung cancer risk by lowering VEGFR1 mRNA expression; the SNP might be a common susceptible locus for both COPD and lung cancer.
Sigma-Aldrich
FLT1 (784-end), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution