Merck
  • Synthesis of quinoxaline 1,4-di-N-oxide analogues and crystal structure of 2-carbomethoxy-3-hydroxyquinoxaline-di-N-oxide.
Synthesis of quinoxaline 1,4-di-N-oxide analogues and crystal structure of 2-carbomethoxy-3-hydroxyquinoxaline-di-N-oxide.
Molecules (Basel, Switzerland) (2011-08-16)
Yingjun Xu, Fanhong Wu, Zhiyi Yao, Minmin Zhang, Sheng Jiang
ABSTRACT
A series of quinoxaline 1,4-di-N-oxide analogues were prepared from benzofurazan N-oxide derivatives and β-diketone ester compounds by the improved Beirut reaction. The structures of the target products were characterized by NMR, MS, IR and elemental analysis measurements, and that of 2-carbomethoxy-3-hydroxyquinoxaline- di-N-oxide was further confirmed by single-crystal X-ray diffraction. Its crystal structure belongs to the monoclinic system, space group C2/c with a = 14.4320 (12) Å, b = 10.7514 (9) Å, c = 13.2728 (11) Å, V = 1958.5 (3) Å 3, Z = 8. The X-ray crystallographic analysis reveals that quinoxaline 1,4-di-N-oxide displays acyloin-endiol tautomerism.
Sigma-Aldrich
Diisopropyl malonate, ReagentPlus®, 99%