Merck
  • Tristetraprolin is involved in the glucocorticoid-mediated interleukin 8 repression.
Tristetraprolin is involved in the glucocorticoid-mediated interleukin 8 repression.
International immunopharmacology (2014-08-12)
Jia-Xin Shi, Jia-Shu Li, Rong Hu, Yi Shi, Xin Su, Xiang-Jun Guo, Xiao-Mei Li
ABSTRACT
Glucocorticoids have been widely used in various inflammatory disorders, and the transcriptional repression of inflammatory mediators has been considered to be the main mechanism of action. However, a previous study showed that dexamethasone inhibited interleukin 8 (IL-8) expression by promoting IL-8 mRNA decay, which implies a posttranscriptional regulation. Nevertheless, by which mechanism dexamethasone destabilized IL-8 mRNA was unclear. Another study indicated that an RNA-binding protein, tristetraprolin (TTP), could be induced by dexamethasone. TTP can bind to AU-rich elements (ARE) in the 3'-untranslated region of target mRNAs and promotes mRNA degradation. So, we speculated that dexamethasone destabilized IL-8 mRNA by upregulating TTP expression. Here, we report that dexamethasone reduced IL-8 expression through destabilizing IL-8 mRNA in human pulmonary microvascular endothelial cells (HPMECs). Dexamethasone stimulation increased TTP mRNA and protein levels. TTP silencing led to mRNA stabilization and protein upregulation of IL-8. These results provide the evidence that the glucocorticoid, in HPMECs, inhibits IL-8 expression through TTP at the posttranscriptional level.
Sigma-Aldrich
MISSION® esiRNA, targeting human UPP1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Upp1
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)