Merck
  • Multicomponent self-sorting of a Pd7 molecular boat and its use in catalytic Knoevenagel condensation.
Multicomponent self-sorting of a Pd7 molecular boat and its use in catalytic Knoevenagel condensation.
Chemical communications (Cambridge, England) (2013-01-09)
Dipak Samanta, Partha Sarathi Mukherjee
ABSTRACT
Unique three-component self-assembly of a cis-blocked 90° Pd(II) acceptor with a mixture of tri- and tetra-imidazole donors led to the self-sorting of a Pd7 molecular boat with an internal nanocavity, which catalyses the Knoevenagel condensation of a series of aromatic aldehydes with 1,3-dimethylbarbituric acid and Meldrum's acid in aqueous media.
Sigma-Aldrich
1,3-Dimethylbarbituric acid, ≥99.0% (T)